EMAIL THIS PAGE TO A FRIEND
The Journal of experimental medicine
Selective control of type I IFN induction by the Rac activator DOCK2 during TLR-mediated plasmacytoid dendritic cell activation.
PMID 20231379
Abstract
Plasmacytoid dendritic cells (pDCs) play a key role in antiviral immunity, but also contribute to the pathogenesis of certain autoimmune diseases, by producing large amounts of type I IFNs. Although activation of pDCs is triggered by engagement of nucleotide-sensing toll-like receptors (TLR) 7 and 9, type I IFN induction additionally requires IkappaB kinase (IKK) alpha-dependent activation of IFN regulatory factor (IRF) 7. However, the signaling pathway mediating IKK-alpha activation is poorly defined. We show that DOCK2, an atypical Rac activator, is essential for TLR7- and TLR9-mediated IFN-alpha induction in pDCs. We found that the exposure of pDCs to nucleic acid ligands induces Rac activation through a TLR-independent and DOCK2-dependent mechanism. Although this Rac activation was dispensable for induction of inflammatory cytokines, phosphorylation of IKK-alpha and nuclear translocation of IRF-7 were impaired in Dock2-deficient pDCs, resulting in selective loss of IFN-alpha induction. Similar results were obtained when a dominant-negative Rac mutant was expressed in wild-type pDCs. Thus, the DOCK2-Rac signaling pathway acts in parallel with TLR engagement to control IKK-alpha activation for type I IFN induction. Owing to its hematopoietic cell-specific expression, DOCK2 may serve as a therapeutic target for type I IFN-related autoimmune diseases.