EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Molecular cloning of a member of a third class of Shaker-family K+ channel genes in mammals.
PMID 2023956
Abstract
In the human proteolipid protein gene, the base sequence of the intronic region 5' to exon 6 was found to be 5'-ctctttcattttcctgcag-3' and not 5'-ctctttt-cattttcctgcag-3' as previously reported.