EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Heterologous expression of mutated eburicol 14alpha-demethylase (CYP51) proteins of Mycosphaerella graminicola to assess effects on azole fungicide sensitivity and intrinsic protein function.
PMID 20305029
Abstract
The recent decrease in the sensitivity of the Western European population of the wheat pathogen Mycosphaerella graminicola to azole fungicides has been associated with the emergence and subsequent spread of mutations in the CYP51 gene, encoding the azole target sterol 14alpha-demethylase. In this study, we have expressed wild-type and mutated M. graminicola CYP51 (MgCYP51) variants in a Saccharomyces cerevisiae mutant carrying a doxycycline-regulatable tetO(7)-CYC promoter controlling native CYP51 expression. We have shown that the wild-type MgCYP51 protein complements the function of the orthologous protein in S. cerevisiae. Mutant MgCYP51 proteins containing amino acid alterations L50S, Y459D, and Y461H and the two-amino-acid deletion DeltaY459/G460, commonly identified in modern M. graminicola populations, have no effect on the capacity of the M. graminicola protein to function in S. cerevisiae. We have also shown that the azole fungicide sensitivities of transformants expressing MgCYP51 variants with these alterations are substantially reduced. Furthermore, we have demonstrated that the I381V substitution, correlated with the recent decline in the effectiveness of azoles, destroys the capacity of MgCYP51 to complement the S. cerevisiae mutant when introduced alone. However, when I381V is combined with changes between residues Y459 and Y461, the function of the M. graminicola protein is partially restored. These findings demonstrate, for the first time for a plant pathogenic fungus, the impacts that naturally occurring CYP51 alterations have on both azole sensitivity and intrinsic protein function. In addition, we also provide functional evidence underlying the order in which CYP51 alterations in the Western European M. graminicola population emerged.