EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Cationic palladium(II) catalysis: C-H activation/Suzuki-Miyaura couplings at room temperature.
PMID 20307066
Abstract
Cationic palladium(II) catalyst realized facile C-H activation of aryl urea with arylboronic acids at room temperature. This reaction is extremely mild to carry out aromatic C-H activations through electrophilic substitution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339768
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
C8H12B2F8N4Pd