EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
In vitro activity of fidaxomicin (OPT-80) tested against contemporary clinical isolates of Staphylococcus spp. and Enterococcus spp.
PMID 20308391