EMAIL THIS PAGE TO A FRIEND
Biological psychiatry
Rapid cerebral amyloid binding by Aβ antibodies infused into β-amyloid precursor protein transgenic mice.
PMID 20359696
Abstract
Passive immunization for the treatment of Alzheimer's disease (AD) was rapidly translated into clinical trials. However, basic mechanisms of AD immunotherapy remain only partially understood. We analyzed the dynamic changes of amyloid-β (Aβ) levels in plasma, brain, and cerebrospinal fluid (CSF) as well as cerebral amyloid binding by Aβ antibody after a single β1-antibody infusion into APP(Swedish) and APP(wildtype) transgenic mice at preplaque and plaque-bearing age. Following intravenous Aβ antibody treatment, plasma Aβ increased rapidly, reaching significantly higher levels in preplaque compared with plaque-bearing mice, whereas cerebral and CSF Aβ remained unchanged. Strikingly, Aβ antibodies exhibited strong cerebral amyloid plaque binding rapidly after intravenous administration in a subset of animals with more severe vascular amyloid. Rapid plasma Aβ increase after Aβ antibody infusion results primarily from stabilization of Aβ. Nevertheless, the smaller plasma Aβ increase in plaque-bearing mice might be of diagnostic use. Importantly, intravenously administered antibodies can rapidly bind to cerebral plaques, potentially facilitated by vascular-amyloid-mediated damage of the blood-brain barrier.