EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Allosteric inhibitors of hepatitis C virus NS5B polymerase thumb domain site II: structure-based design and synthesis of new templates.
PMID 20363140
Abstract
Chronic hepatitis C virus (HCV) infections are a significant medical problem worldwide. The NS5B Polymerase of HCV plays a central role in virus replication and is a prime target for the discovery of new treatment options. We recently disclosed 1H-benzo[de]isoquinoline-1,3(2H)-diones as allosteric inhibitors of NS5B Polymerase. Structural and SAR information guided us in the modification of the core structure leading to new templates with improved activity and toxicity/activity window.