EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Azetidinone-isothiazolidinones: stereoselective synthesis and antibacterial evaluation of new monocyclic beta-lactams.
PMID 20382539
Abstract
A simple and efficient procedure for the stereoselective synthesis of new azetidinone-isothiazolidinones has been developed. New compounds were tested in vitro on a panel of Gram-positive and Gram-negative bacterial pathogens, some of them showing weak antibacterial activity.