EMAIL THIS PAGE TO A FRIEND
Cancer research
A unique pharmacophore for activation of the nuclear orphan receptor Nur77 in vivo and in vitro.
PMID 20388790
Abstract
Nur77 is a steroid orphan receptor that plays a critical role in regulating proliferation, differentiation, and apoptosis, including acting as a switch for Bcl-2 function. We previously reported that the octaketide cytosporone B (Csn-B) is a natural agonist for Nur77. In this study, we synthesized a series of Csn-B analogues and performed a structure-activity analysis that suggested criteria for the development of a unique pharmacophore to activate Nur77. The components of the pharmacophore necessary for binding Nur77 included the benzene ring, the phenolic hydroxyl group, and the acyl chain of the Csn-B scaffold, whereas the key feature for activating the biological function of Nur77 was the ester group. Csn-B analogues that bound Nur77 tightly not only stimulated its transactivation activity but also initiated mitochondrial apoptosis by means of novel cross-talk between Nur77 and BRE, an antiapoptotic protein regulated at the transcriptional level. Notably, the derivative n-amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate exhibited greater antitumor activity in vivo than its parent compounds, highlighting particular interest in this compound. Our findings describe a pathway for rational design of Csn-B-derived Nur77 agonists as a new class of potent and effective antitumor agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0987
n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate, ≥98% (HPLC)
C22H34O5