EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Probe-based 3-D nanolithography using self-amplified depolymerization polymers.
PMID 20419710
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900029
Polyphthalaldehyde, Linear