EMAIL THIS PAGE TO A FRIEND
Epigenetics & chromatin
HP1gamma function is required for male germ cell survival and spermatogenesis.
PMID 20423503
Abstract
HP1 proteins are conserved components of eukaryotic constitutive heterochromatin. In mammals, there are three genes that encode HP1-like proteins, termed HP1alpha, HP1beta and HP1gamma, which have a high degree of homology This paper describes for the first time, to our knowledge, the physiological function of HP1gamma using a gene-targeted mouse. While targeting the Cbx3 gene (encoding the HP1gamma protein) with a conditional targeting vector, we generated a hypomorphic allele (Cbx3hypo), which resulted in much reduced (barely detectable) levels of HP1gamma protein. Homozygotes for the hypomorphic allele (Cbx3hypo/hypo) are rare, with only 1% of Cbx3hypo/hypo animals reaching adulthood. Adult males exhibit a severe hypogonadism that is associated with a loss of germ cells, with some seminiferous tubules retaining only the supporting Sertoli cells (Sertoli cell-only phenotype). The percentage of seminiferous tubules that are positive for L1 ORF1 protein (ORF1p) in Cbx3hypo/hypo testes is greater than that for wild-type testes, indicating that L1 retrotransposon silencing is reversed, leading to ectopic expression of ORF1p in Cbx3hypo/hypo germ cells. The Cbx3 gene product (the HP1gamma protein) has a non-redundant function during spermatogenesis that cannot be compensated for by the other two HP1 isotypes. The Cbx3hypo/hypo spermatogenesis defect is similar to that found in Miwi2 and Dnmt3L mutants. The Cbx3 gene-targeted mice generated in this study provide an appropriate model for the study of HP1gamma in transposon silencing and parental imprinting.