EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Novel bisstyryl derivatives of bakuchiol: targeting oral cavity pathogens.
PMID 20427099
Abstract
Novel bisstyryl derivatives of bakuchiol using Heck coupling reaction as the key step were synthesized and screened against a panel of six oral cavity pathogens for their antimicrobial activity. Four compounds (9-12) showed two to fourfold and four to eightfold better activity (MIC 0.25-16 microg/ml) than bakuchiol and triclosan respectively. These compounds effectively inhibit the biofilm formation of single and multiple species at 2 - 8 x MICs. 4- and 4'-Hydroxy/methoxy styryl moieties of the bakuchiol derivatives play a pivotal role towards the activity as established in the SAR studies. Mechanism of action studies revealed microbial membrane structure disruption as the probable mode of action of these compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68612
Bakuchiol, analytical standard
C18H24O