EMAIL THIS PAGE TO A FRIEND
Biochemistry
Alkaline phosphatase: affinity chromatography and inhibition by phosphonic acids.
PMID 204333
Abstract
Five phosphonic acid derivatives were synthesized, coupled to agarose, and tested for affinity chromatographic binding of alkaline phosphatase from bovine intestine. Agarose coupled to L-histidyldiazobenzylphosphonic acid was found to be a highly effective adsorbent. In order to understand the large differences in binding capacity observed with derivatized agaroses, inhibition of alkaline phosphatase by phosphonic acid ligands, and related phosphonic acids, was measured. The results of affinity chromatography and inhibition studies were in good agreement, demonstrating that phosphonic acids with large aromatic/hydrophobic, carboxylate substituents bind strongly and competitively to the enzyme active site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6758 L-Histidyldiazobenzylphosphonic acid-Agarose, saline suspension