EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Semisynthesis and pharmacological activities of thyroxine analogs: Development of new angiogenesis modulators.
PMID 20434332
Abstract
Novel thyroxine analogs with hindered phenol, amino and carboxylic acid groups have been synthesized and the effects of the synthesized compounds on angiogenesis using the chick chorioallantoic membrane and mouse matrigel models have been tested. Pharmacological profiles revealed that thyroxine tolerates numerous modifications on the amino group and remains active. These results provide the rationale for the selection of a novel thyroxine nanoparticle precursor.