EMAIL THIS PAGE TO A FRIEND
Biochemistry
Multiple isotope effects with alternative dinucleotide substrates as a probe of the malic enzyme reaction.
PMID 2043615
Abstract
Deuterium isotope effects and 13C isotope effects with deuterium- and protium-labeled malate have been obtained for both NAD- and NADP-malic enzymes by using a variety of alternative dinucleotide substrates. With nicotinamide-containing dinucleotides as the oxidizing substrate, the 13C effect decreases when deuterated malate is the substrate compared to the value obtained with protium-labeled malate. These data are consistent with a stepwise chemical mechanism in which hydride transfer precedes decarboxylation of the oxalacetate intermediate as previously proposed [Hermes, J. D., Roeske, C. A., O'Leary, M. H., & Cleland, W. W. (1982) Biochemistry 21, 5106]. When dinucleotide substrates such as thio-NAD, 3-acetylpyridine adenine dinucleotide, and 3-pyridinealdehyde adenine dinucleotide that contain modified nicotinamide rings are used, the 13C effect increases when deuterated malate is the substrate compared to the value obtained with protium-labeled malate. These data, at face value, are consistent with a change in mechanism from stepwise to concerted for the oxidative decarboxylation portion of the mechanism. However, the increase in the deuterium isotope effect from 1.5 to 3 with a concomitant decrease in the 13C isotope effect from 1.034 to 1.003 as the dinucleotide substrate is changed suggests that the reaction may still be stepwise with the non-nicotinamide dinucleotides. A more likely explanation is that a beta-secondary 13C isotope effect accompanies hydride transfer as a result of hyperconjugation of the beta-carboxyl of malate as the transition state for the hydride transfer step is approached.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7375
Thionicotinamide adenine dinucleotide, ≥90%
C21H27N7O13P2S