EMAIL THIS PAGE TO A FRIEND
Pathology
Theranostic and prognostic biomarkers: genomic applications in urological malignancies.
PMID 20438413
Abstract
Compared to other solid tumours such as breast, colon, and lung, the current clinical management of urological malignancies is lagging behind in terms of utilisation of clinically robust molecular tests that can identify patients that are more likely to respond to a given targeted agent, or even those in need of a more aggressive treatment approach based on well-validated molecular prognosticators. Several promising biomarkers for detection, prognosis, and targeted therapeutics are now under evaluation. The following review discusses some of the candidate biomarkers that may soon make their transition into clinically applicable assays in urological oncology patients.