EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Agonist/antagonist modulation in a series of 2-aryl benzimidazole H4 receptor ligands.
PMID 20452213
Abstract
The present work details the transformation of a series of human histamine H(4) agonists into potent functional antagonists. Replacement of the aminopyrrolidine diamine functionality with a 5,6-fused pyrrolopiperidine ring system led to an antagonist. The dissection of this fused diamine led to the eventual replacement with heterocycles. The incorporation of histamine as the terminal amine led to a very potent and selective histamine H(4) agonist; whereas incorporation of the constrained histamine analog, spinacamine, modulated the functional activity to give a partial agonist. In two separate series, we demonstrate that constraining the terminal amino portion modulated the spectrum of functional activity of histamine H(4) ligands.