EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Mesoporous silica nanoparticles facilitate delivery of siRNA to shutdown signaling pathways in mammalian cells.
PMID 20461725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
435716
3-(Trihydroxysilyl)propyl methylphosphonate, monosodium salt solution, 50 wt. % in H2O
C4H12NaO6PSi