EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Design and synthesis of 2,4-disubstituted polyhydroquinolines as prospective antihyperglycemic and lipid modulating agents.
PMID 20471838
Abstract
A series of 2,4-disubstituted polyhydroquinoline were synthesized and evaluated for their in vivo antihyperglycemic as well as antidyslipidemic activities. Several synthesized compounds have exhibited promising in vivo antihyperglycemic in SLM, STZ-S, and db/db mice model along with significant lipid and TG modulating activity. All these compounds were evaluated in various in vitro models of diabetes to know the possible mechanism of their antihyperglycemic action. Interestingly, compounds 3a-r (diaryl substitution) have exhibited promising protein-tyrosine phosphatase 1B (PTP1B) inhibitory activity whereas, compounds 5a-d (acid substituted) have shown significant glycogen phosphorylase activity.