EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
A novel polymyxin derivative that lacks the fatty acid tail and carries only three positive charges has strong synergism with agents excluded by the intact outer membrane.
PMID 20479195
Abstract
Polymyxins are cationic lipopeptides (five cationic charges) and the last resort for the treatment of serious Gram-negative infections caused by multiresistant strains. NAB741 has a cyclic peptide portion identical to that of polymyxin B but carries in the linear peptide portion a threonyl-D-serinyl residue (no cationic charges) instead of the diaminobutyryl-threonyl-diaminobutyryl residue (two cationic charges). At the N terminus of the peptide, NAB741 carries an acetyl group instead of a mixture of methyl octanoyl and methyl heptanoyl residues. NAB741 sensitized Escherichia coli, Klebsiella pneumoniae, Enterobacter cloacae, and Acinetobacter baumannii to antibiotics against which the intact outer membrane is an effective permeability barrier. When tested by using Etest strips on plates containing increasing concentrations of NAB741, the fractional inhibition concentration index (FICI) of the combination of NAB741 with rifampin ranged from
Related Materials