EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
New pyrazolo[1,5a]pyrimidines as orally active inhibitors of Lck.
PMID 20483608
Abstract
A novel series of pyrazolo[1,5a]pyrimidines was optimized to target lymphocyte-specific kinase (Lck). An efficient synthetic route was developed and SAR studies toward activity and selectivity are described, leading to Lck inhibitors with enzymatic, cellular and in vivo potency.