EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis of novel beta-lactam fused spiroisoxazolidine chromanones and tetralones as potent antimicrobial agent for human and plant pathogens.
PMID 20483609
Abstract
Synthesis of novel beta-lactam fused spiroisoxazolidine chromanones and tetralones ring systems has been achieved by intermolecular 1,3-dipolar cycloaddition reaction of bicyclic nitrone with unusual dipolarophiles, arylidene chromanones/tetralones under different reaction conditions. The synthesized compounds were evaluated for antimicrobial activities. It was observed that two of the synthesized compounds exhibited relatively good antibacterial and antifungal activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45368
Carbendazim, PESTANAL®, analytical standard
C9H9N3O2