EMAIL THIS PAGE TO A FRIEND
Gene therapy
Mifepristone increases gamma-retroviral infection efficiency by enhancing the integration of virus into the genome of infected cells.
PMID 20485384
Abstract
Gamma-retroviruses are commonly used to deliver genes to cells. Previously, we demonstrated that the synthetic anti-glucocorticoid and anti-progestin agent, mifepristone, increased gamma-retroviral infection efficiency in different target cells, independent of viral titer. In this study, we examine how this occurs. We studied the effect of mifepristone on different steps of viral infection (viral entry, viral survival, viral DNA synthesis and retrovirus integration into the host genome) in three distinct retroviral backbones using different virus recognition receptors. We also tested the potential role of glucocorticoid and progesterone receptors in mediating mifepristone's ability to increase gamma-retroviral infectivity. We show that mifepristone increases gamma-retroviral infection efficiency by facilitating viral integration into the host genome and that this effect seems to be due to mifepristone's anti-glucocorticoid, but not its anti-progestin, activity. These results suggest that inhibition of the glucocorticoid receptor enhances retroviral integration into the host genome and indicates that cells may have a natural protection again retroviral infection that may be reduced by glucocorticoid receptor antagonists.
Related Materials