EMAIL THIS PAGE TO A FRIEND
The journal of sexual medicine
Expression of cyclic AMP-dependent protein kinase isoforms in human cavernous arteries: functional significance and relation to phosphodiesterase type 4.
PMID 20487244
Abstract
The cyclic adenosine monophosphate-dependent protein kinase (cAK) is considered a key protein in the control of smooth muscle tone in the cardiovascular system. There is evidence that erectile dysfunction might be linked to systemic vascular disorders and arterial insufficiency, subsequently resulting in structural changes in the penile tissue. The expression and significance of cAK in human cavernous arteries (HCA) have not been evaluated. To evaluate the expression of cAK isoforms in HCA and examine the role of cAK in the cyclic adenosine monophosphate (cAMP)- and cyclic guanosine monophosphate (cGMP)-mediated control of penile vascular smooth muscle. The expression and distribution of phosphodiesterase type 4 (PDE4) and cAK isoforms in sections of HCA were investigated by means of immunohistochemistry and Western blot analysis. The effects of the cAK inhibitor Rp-8-CPT-cAMPS on the relaxation of isolated preparations of HCA (diameter > 100 µm) induced by rolipram, sildenafil, tadalafil, and vardenafil were studied using the organ bath technique. Investigate the expression of cAK in relation to α-actin and PDE4 in HCA and evaluate the effects of an inhibition of cAK on the relaxation induced by inhibitors of PDE4 and PDE5 of isolated penile arteries. Immunosignals specific for cAKIα, IIα, and IIβ were observed within the wall of HCA. Double stainings revealed colocalization of cAK with α-actin and PDE4. The expression of cAK isoforms was confirmed by Western blot analysis. The reversion of tension induced by inhibitors of PDE4 and PDE5 of isolated penile vascular tissue were attenuated significantly by Rp-8-CPT-cAMPS. Our results demonstrate the expression of cAK isoforms in the smooth musculature of HCA and its colocalization with PDE4. A significant role for cAK in the regulation mediated by cAMP and cGMP of vascular smooth muscle tone in HCA can also be assumed.