EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
In silico characterization of cytisinoids docked into an acetylcholine binding protein.
PMID 20493692
Abstract
Homology models of nicotinic acetylcholine receptors (nAChRs) suggest that subtype specificity is due to non-conserved residues in the complementary subunit of the ligand-binding pocket. Cytisine and its derivatives generally show a strong preference for heteromeric alpha4beta2* nAChRs over the homomeric alpha7 subtype, and the structural modifications studied do not cause large changes in their nAChR subtype selectivity. In the present work we docked cytisine, N-methylcytisine, and several pyridone ring-substituted cytisinoids into the crystallographic structure of the Lymnaea stagnalis acetylcholine binding protein (AChBP) co-crystallized with nicotine (1UW6). The graphical analysis of the best poses showed that cytisinoids have weak interactions with the side chains of the non-conserved amino acids in the complementary subunit justifying the use of PDB 1UWB as a surrogate for nAChR. Furthermore, we found a high correlation (R(2)=0.96) between the experimental pIC(50) values at alpha4beta2* nAChR and docking energy (S) of the best cytisinoid poses within the AChBP. Due to the quality of the correlation we suggest that this equation might be used as a predictive model to propose new cytisine-derived nAChRs ligands. Our docking results also suggest that further structural modifications of these cytisinoids will not greatly alter their alpha4beta2*/alpha7 selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2899
Cytisine, ≥99%, powder
C11H14N2O