EMAIL THIS PAGE TO A FRIEND
Life sciences
Effect of simvastatin on Kruppel-like factor2, endothelial nitric oxide synthase and thrombomodulin expression in endothelial cells under shear stress.
PMID 20493886
Abstract
Statins (3-hydroxy-3-methylglutaryl coenzyme A reductase inhibitors) and fluid wall shear stress have been reported to modulate the expression of genes related to inflammation, blood coagulation, thrombosis, and vascular constriction in cultured endothelial cells. In this study, we investigated the combined effect of laminar shear stress (LSS) and statins on endothelial cell gene expression. Kruppel-like factor 2 (KLF2), endothelial nitric oxide synthase (eNOS), and thrombomodulin (TM) mRNA and protein expression were evaluated in human abdominal aortic endothelial cells (HAAEC) treated with simvastatin (0.1, 1 or 10 microM) at various levels of LSS (0, 1.25, 12.5 or 25 dynes/cm(2)). As expected, simvastatin and LSS separately enhanced KLF2, eNOS, and TM mRNA expressions. The combination of simvastatin and LSS resulted in significantly higher mRNA levels of all three genes compared to cells treated with LSS only. The highest KLF2, eNOS, and TM mRNA levels were detected at 10 microM simvastatin and 25 dynes/cm(2). Under these conditions, eNOS and TM protein levels were also elevated. Combining LSS and simvastatin produced an overall additive increase in KLF2, eNOS, and TM mRNA. Treatment of the endothelial cells with 10 microM simvastatin and 200 microM mevalonate completely eliminated the effect of simvastatin. Our results suggest an additive increase in KLF2, eNOS, and TM expressions when simvastatin and LSS are combined. These results may help to explain the proposed non-lipid lowering benefits of statins observed in the clinic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6196
Simvastatin, ≥97% (HPLC), solid
C25H38O5