EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, synthesis, and antibacterial activity of 2,5-dihydropyrrole formyl hydroxyamino derivatives as novel peptide deformylase inhibitors.
PMID 20510610
Abstract
The synthesis and antibacterial activity of 2,5-dihydropyrrole formyl hydroxyamino derivatives are reported. The antibacterial activities of these derivatives were evaluated, and some of these derivatives showed better in vitro antibacterial activity than existing drugs, including penicillin, ciprofloxacin, vancomycin, and linezolid.