EMAIL THIS PAGE TO A FRIEND
Nature structural & molecular biology
Structural basis of G-tract recognition and encaging by hnRNP F quasi-RRMs.
PMID 20526337
Abstract
The heterogeneous nuclear ribonucleoprotein (hnRNP) F is involved in the regulation of mRNA metabolism by specifically recognizing G-tract RNA sequences. We have determined the solution structures of the three quasi-RNA-recognition motifs (qRRMs) of hnRNP F in complex with G-tract RNA. These structures show that qRRMs bind RNA in a very unusual manner, with the G-tract 'encaged', making the qRRM a novel RNA binding domain. We defined a consensus signature sequence for qRRMs and identified other human qRRM-containing proteins that also specifically recognize G-tract RNAs. Our structures explain how qRRMs can sequester G-tracts, maintaining them in a single-stranded conformation. We also show that isolated qRRMs of hnRNP F are sufficient to regulate the alternative splicing of the Bcl-x pre-mRNA, suggesting that hnRNP F would act by remodeling RNA secondary and tertiary structures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10203 CompoZr® Knockout ZFN Kit, Human HNRNPF (NM_001098204)