EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
4-Substituted-7-N-alkyl-N-acetyl 2-aminobenzothiazole amides: drug-like and non-xanthine based A2B adenosine receptor antagonists.
PMID 20541935
Abstract
7-N-Acetamide-4-methoxy-2-aminobenzothiazole 4-fluorobenzamide (compound 1) was chosen as a drug-like and non-xanthine based starting point for the discovery of A(2B) receptor antagonists because of its slight selectivity against A(1) and A(2A) receptors and modest A(2B) potency. SAR exploration of compound 1 described herein included modifications to the 7-N-acetamide group, substitution of the 4-methoxy group by halogens as well as replacement of the p-flouro-benzamide side chain. This work culminated in the identification of compound 37 with excellent A(2B) potency, modest selectivity versus A(2A) and A(1) receptors, and good rodent PK properties.