EMAIL THIS PAGE TO A FRIEND
Cell research
Coordinated regulation of active and repressive histone methylations by a dual-specificity histone demethylase ceKDM7A from Caenorhabditis elegans.
PMID 20567262
Abstract
H3K9me2 and H3K27me2 are important epigenetic marks associated with transcription repression, while H3K4me3 is associated with transcription activation. It has been shown that active and repressive histone methylations distribute in a mutually exclusive manner, but the underlying mechanism was poorly understood. Here we identified ceKDM7A, a PHD (plant homeodomain)- and JmjC domain-containing protein, as a histone demethylase specific for H3K9me2 and H3K27me2. We further demonstrated that the PHD domain of ceKDM7A bound H3K4me3 and H3K4me3 co-localized with ceKDM7A at the genome-wide level. Disruption of the PHD domain binding to H3K4me3 reduced the demethylase activity in vivo, and loss of ceKDM7A reduced the expression of its associated target genes. These results indicate that ceKDM7A is recruited to the promoter to demethylate H3K9me2 and H3K27me2 and activate gene expression through the binding of the PHD domain to H3K4me3. Thus, our study identifies a dual-specificity histone demethylase and provides novel insights into the regulation of histone methylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-585
Anti-dimethyl-Histone H3 (Arg2) Antibody, Anti-dimethyl-Histone H3 (Arg2) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2) & has been validated in WB, PIA.
07-452
Anti-dimethyl-Histone H3 (Lys27) Antibody, Use Anti-dimethyl-Histone H3 (Lys27) Antibody (rabbit polyclonal antibody) validated in Mplex, PIA, WB, ChIP-seq, DB to detect dimethyl-Histone H3 (Lys27) also known as H3K27me2, Histone H3 (di methyl K27).
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
07-448
Anti-monomethyl-Histone H3 (Lys27) Antibody, Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in ICC, WB, ChIP, ChIP-seq.
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ChIP, ChIP-seq, ICC, IP.
05-745R
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, rabbit monoclonal, Use Anti-trimethyl-Histone H3 (Lys4) Antibody, clone 15-10C-E4, Recombinant (rabbit monoclonal antibody) proven in WB, ChIP, ChIP-seq, ELISA, PIA, Mplex, DB to detect trimethyl-Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4).
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
12-570
Monomethyl-Histone H3 (Lys36) Peptide, biotin conjugate, Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.