EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Synthesis of colloidal silica nanoparticles of a tunable mesopore size and their application to the adsorption of biomolecules.
PMID 20570275
Abstract
A synthetic method for the fabrication of colloidal mesoporous silica nanoparticles of a tunable mesopore size is presented. These nanoparticles form stable colloidal suspensions, have narrow particle size distributions in the 50-300nm range and exhibit a framework-confined porosity made up of large mesopores with a diameter of up to 9nm. The size of the mesopores was enlarged by means of two different swelling agents (SA), i.e. 1,3,5-trimethylbenzene (TMB) and N,N-dimethylhexadecylamine (DMHA). Colloidal mesoporous silica nanoparticles exhibit a strong affinity for the adsorption of hemoglobin (Hb) and the immobilization of lysozyme (Lz). These nanoparticles can adsorb up to 2080mgHbg(-1) support and 690mgLzg(-1) support. Moreover, the adsorption of these biomolecules on silica nanoparticles occurs very quickly and most of what is adsorbed is accommodated on the external surface area of the silica nanoparticles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40460
N,N-Dimethylhexadecylamine, technical, ≥95% (GC)
C18H39N