EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
Iron traffics in circulation bound to a siderocalin (Ngal)-catechol complex.
PMID 20581821
Abstract
The lipocalins are secreted proteins that bind small organic molecules. Scn-Ngal (also known as neutrophil gelatinase associated lipocalin, siderocalin, lipocalin 2) sequesters bacterial iron chelators, called siderophores, and consequently blocks bacterial growth. However, Scn-Ngal is also prominently expressed in aseptic diseases, implying that it binds additional ligands and serves additional functions. Using chemical screens, crystallography and fluorescence methods, we report that Scn-Ngal binds iron together with a small metabolic product called catechol. The formation of the complex blocked the reactivity of iron and permitted its transport once introduced into circulation in vivo. Scn-Ngal then recycled its iron in endosomes by a pH-sensitive mechanism. As catechols derive from bacterial and mammalian metabolism of dietary compounds, the Scn-Ngal-catechol-Fe(III) complex represents an unforeseen microbial-host interaction, which mimics Scn-Ngal-siderophore interactions but instead traffics iron in aseptic tissues. These results identify an endogenous siderophore, which may link the disparate roles of Scn-Ngal in different diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M34200
4-Methylcatechol, ≥95%
C7H8O2