EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Plasmid-mediated resistance to cephalosporins in Salmonella enterica serovar Typhi.
PMID 20585124
Abstract
This study characterized a cephalosporin-resistant Salmonella enterica serovar Typhi isolate. The organism possessed a plasmid encoding the CTX-M-15 extended-spectrum-beta-lactamase. This plasmid is the determinant for the phenotype of cephalosporin resistance and is transferrable among Enterobacteriaceae.