EMAIL THIS PAGE TO A FRIEND
Optics express
Broadband NIR photoluminescence from Bi-doped Ba2P2O7 crystals: insights into the nature of NIR-emitting Bismuth centers.
PMID 20588414
Abstract
We report on a novel type of Bi-doped crystal that exhibits ultrabroadband photoluminescence in the near infrared (NIR). Emission centers can be generated and degenerated reversibly by annealing the material in CO atmosphere and air, respectively, indicating that emission is related to the presence of Bi-species in low valence states. Correlating static and dynamic excitation and emission data with the size and charge of available lattice sites suggests that two types of Bi(0)-species, each located on one of the two available Ba(2+) lattice sites, are responsible for NIR photoemission. This is further confirmed by the absence of NIR emission in polycrystalline Ca(2)P(2)O(7):Bi and Sr(2)P(2)O(7):Bi. Excitation is assigned to transitions between the doubly degenerated ground state (4)S(3/2) and the degenerated excited levels (2)D(3/2), (2)D(5/2) and (2)P(1/2), respectively. NIR emission is attributed to (2)D(3/2)?(4)S(3/2). The NIR emission center can coexist with Bi(2+) species. Then, also Bi(2+) is accommodated on one of the two Ba(2+)-sites. Energy transfer between Bi(2+) ions occurs within a critical distance of 25.9 A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
772658
Barium pyrophosphate, powder, -200 mesh, 99.9% trace metals basis
Ba2P2O7