EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and CYP24A1 inhibitory activity of N-(2-(1H-imidazol-1-yl)-2-phenylethyl)arylamides.
PMID 20594862
Abstract
A series of N-(2-(1H-imidazol-1-yl)-2-phenylethyl)arylamides were prepared, using an efficient three- to five-step synthesis, and evaluated for their inhibitory activity against human cytochrome P450C24A1 (CYP24A1) hydroxylase. Inhibition ranged from IC50 0.3-72 microM compared with the standard ketoconazole IC50 0.52 microM, with the styryl derivative (11c) displaying enhanced activity (IC50=0.3 microM) compared with the standard, providing a useful preliminary lead for drug development.