EMAIL THIS PAGE TO A FRIEND
Cellular signalling
TGF-beta1 suppresses IL-6-induced STAT3 activation through regulation of Jak2 expression in prostate epithelial cells.
PMID 20603212
Abstract
Chronic inflammation plays an important role in the initiation and progression of various human diseases including benign prostatic hyperplasia or prostate cancer. Here we show that the proinflammatory cytokine interleukin-6 (IL-6) has prosurvival effects and chronically activates the Jak2/STAT3 signalling pathway in a model of benign prostatic hyperplasia (BPH-1). We demonstrate that the antiinflammatory cytokine transforming growth factor-beta1 (TGF-beta1), which also permanently activates its canonical signalling pathway through SMAD proteins in BPH-1 cells, modifies the effects of IL-6 on cell proliferation. Importantly, TGF-beta1 inhibits IL-6 signal transduction by decreasing the phosphorylation levels of STAT3. This effect is associated with decreased expression of Jak2 at both mRNA and protein levels. Moreover, we showed that TGF-beta1 inhibits IL-6-induced expression of the cancer-associated gene MUC1. These observations demonstrated a novel interaction between TGF-beta1 and IL-6 signalling and suggested another mechanism of how defects in TGF-beta signalling, frequently associated with prostate pathologies, can contribute to the disruption of tissue homeostasis.