EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Design, synthesis, and anticonvulsant activity of new N-Mannich bases derived from spirosuccinimides and spirohydantoins.
PMID 20638856
Abstract
The synthesis and anticonvulsant properties of new N-Mannich bases of [7,8-f]benzo-2-aza-spiro[4.5]decane-1,3-diones (5a-h) and [7,8-f]benzo-1,3-diaza-spiro[4.5]decane-2,4-diones (7a-h) were described. Initial anticonvulsant screening was performed using intraperitoneal (ip) maximal electroshock (MES) and subcutaneous pentylenetetrazole (scPTZ) seizures tests. The neurotoxicity was determined applying the rotarod test. The majority of compounds were effective in the MES or/and scPTZ screen. The quantitative studies showed that several molecules were more potent than phenytoin, used as reference drug. Selected derivatives were screened in the 6-Hz test and also assessed for potential activity against nerve agents using the Pilocarpine Induced Status Prevention model. To explain the possible mechanism of anticonvulsant action, for chosen active derivatives, their influence on voltage-dependent Na(+) channel were tested in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9381
Succinimide
C4H5NO2