EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Study of inhibitors of the PGH synthases for which potency is regulated by the redox state of the enzymes.
PMID 20645166
Abstract
Aspirin, salicylic acid, and acetaminophen are examples of drugs that inhibit the PGH synthases (cyclooxygenases) with potencies that vary remarkably between different cells. This results from the fact that the inhibitory action of these drugs is regulated by redox cycling of the enzymes, which is determined by the concentration of hydroperoxides in the enzyme proximity. The potency of these drugs is greatest in the presence of low concentrations of hydroperoxides. Accordingly, analysis of the action of these inhibitors requires consideration of assay conditions that take into account the factors that control hydroperoxide concentrations, both in studies with purified enzymes and in cells.
Related Materials