EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Neuroprotective mechanisms of brain-derived neurotrophic factor against 3-nitropropionic acid toxicity: therapeutic implications for Huntington's disease.
PMID 20649532
Abstract
3-Nitropropionic acid (3-NP) is an irreversible inhibitor of mitochondrial succinate dehydrogenase that has been used to explore the molecular mechanisms of cell death associated with mitochondrial dysfunction and neurodegeneration for Huntington's disease (HD). Brain-derived neurotrophic factor (BDNF) is a neurotrophin that may regulate neuronal survival and differentiation. Experimental evidence derived from both clinical as well as basic research suggests a close association between BDNF deficiency and HD pathogenesis. In this review, we focus on recent progress in the molecular mechanisms responsible for the BDNF-mediated neuroprotective effects against mitochondrial dysfunction induced by 3-NP. Delineation of BDNF-mediated neuroprotective actions against 3-NP toxicity may add in the development of therapeutic intervention for HD where mitochondrial dysfunction is known to play a crucial role in pathogenesis of this devastating disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5636
3-Nitropropionic acid, ≥97%
C3H5NO4