EMAIL THIS PAGE TO A FRIEND
Anticancer research
Suppression by L-methionine of cell cycle progression in LNCaP and MCF-7 cells but not benign cells.
PMID 20651330
Abstract
Methionine inhibits proliferation of breast and prostate cancer cells. This study aimed to determine cell cycle effects of methionine and selectivity for cancer cells. MCF-7 (breast), LNCaP (prostate), and LS-174 (colon) cancer cells (wild-type p53), DU-145 (prostate) and SW480 (colon) cancer cells (mutated p53), and immortalized, non-tumorigenic MCF-10A (breast), BPH-1 (prostate), and NCM-460 (colon) epithelial cells were used. Cell cycle effects were assessed by flow cytometry and cell cycle-related gene expression by microarray analysis and QRT-PCR. L-Methionine at 5 mg/ml for 72 hours (non-apoptotic) arrested cell cycle in LNCaP, DU145, and MCF-7 cells, but not in untransformed cells, nor in LS-174 cells. LNCaP and MCF-7 cells were arrested at G(1), but DU-145 at S. Methionine up-regulated CDKIs and down-regulated CDKs. L-Methionine selectively inhibits proliferation of breast and prostate cancer cells, but not non-tumorigenic cells, and may thus have therapeutic benefits. p53 status appeared to determine the cell cycle stage at which methionine acts.