EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
An efficient route to xanthine based A(2A) adenosine receptor antagonists and functional derivatives.
PMID 20652178
Abstract
A one-pot route to 8-substituted xanthines has been developed from 5,6-diaminouracils and carboxaldehydes. Yields are good and the process applicable to a range of substrates including a family of A(2A) adenosine receptor antagonists. A new route to the KW-6002 family of antagonists is presented including a pro-drug variant, and application to related image contrast agents developed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
680524
2-Chloro-5-(trifluoromethyl)phenylboronic acid
C7H5BClF3O2
SML0422
Istradefylline, ≥98% (HPLC)
C20H24N4O4