EMAIL THIS PAGE TO A FRIEND
Neural development
Induction of early Purkinje cell dendritic differentiation by thyroid hormone requires RORα.
PMID 20663205
Abstract
The active form (T3) of thyroid hormone (TH) controls critical aspects of cerebellar development, such as migration of postmitotic neurons and terminal dendritic differentiation of Purkinje cells. The effects of T3 on early dendritic differentiation are poorly understood. In this study, we have analyzed the influence of T3 on the progression of the early steps of Purkinje cell dendritic differentiation in postnatal day 0 organotypic cerebellar cultures. These steps include, successively, regression of immature neuritic processes, a stellate cell stage, and the extension of several long and mature perisomatic protrusions before the growth of the ultimate dendritic tree. We also studied the involvement of RORalpha, a nuclear receptor controlling early Purkinje cell dendritic differentiation. We show that T3 treatment leads to an accelerated progression of the early steps of dendritic differentiation in culture, together with an increased expression of RORalpha (mRNA and protein) in both Purkinje cells and interneurons. Finally, we show that T3 failed to promote early dendritic differentiation in staggerer RORalpha-deficient Purkinje cells. Our results demonstrate that T3 action on the early Purkinje cell dendritic differentiation process is mediated by RORalpha.