EMAIL THIS PAGE TO A FRIEND
Farmacia hospitalaria : organo oficial de expresion cientifica de la Sociedad Espanola de Farmacia Hospitalaria
Topical endotracheal mitomycin C as a complementary treatment for endoscopic treatment of recurrent laryngotracheal stenosis.
PMID 20663704
Abstract
To describe the preparation of topical endotracheal mitomycin C and the clinical outcomes of four patients with recurrent and severe laryngotracheal stenosis (LTS) treated with adjuvant topical mitomycin C. Literature review to determine the concentration and method of preparation of topical mitomycin C for endotracheal use. Review of clinical histories. We established a concentration of 0.4 mg/ml topical mitomycin C for the treatment of laryngotracheal stenosis. In the treated cases, we applied a 0.4 mg/ml solution to the wound site following laser surgery and dilatation with bronchoscope. Three patients remain asymptomatic from a respiratory perspective, and treatment failed in one case. LTS treatment is complex due to the continuous development of granulation tissue and fibrosis following injury to the airways. Topical mitomycin C seems to be the ideal adjuvant agent thanks to its powerful antifibrotic effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000378 Mitomycin, European Pharmacopoeia (EP) Reference Standard
C15H18N4O5