EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Ammonia-borane and related compounds as dihydrogen sources.
PMID 20672860
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
737488
Borane ethylenediamine complex, 96%
C2H14B2N2