EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Membrane efflux and influx modulate both multidrug resistance and virulence of Klebsiella pneumoniae in a Caenorhabditis elegans model.
PMID 20679507
Abstract
Cross-resistance to cefoxitin (FOX), chloramphenicol (CMP), and quinolones (nalidixic acid [NAL]) related to a putative efflux system overexpression has recently been reported for Klebsiella pneumoniae. The potential impact of this multidrug resistance (MDR) on the virulence of K. pneumoniae was evaluated in the Caenorhabditis elegans model. For 2 of the 3 MDR clinical isolates studied, a significant increase in acrB transcription was found in comparison with their antibiotic-susceptible revertants. ATCC 138821 and MDR, revertant, and derivative strains with altered porin expression were studied. Strains proved or suspected to overexpress an efflux system were significantly more virulent than the ATCC and revertant strains (time to kill 50% of nematodes [LT(50)] in days: 3.4 to 3.8 ± 0.2 versus 4.1 to 4.4 ± 0.3, P < 0.001). Inversely, strains with altered porin expression were significantly less virulent, independently of the expression level of efflux system (LT(50) = 5.4 to 5.6 ± 0.2, P < 0.001). Altered porin expression did not change MICs of CMP and NAL but did those of FOX (4 to 16× MIC) and ertapenem (16 to 64× MIC). The strains with a normally or an overexpressed efflux system that received the β-lactamase CTX-M-15 became more widely resistant without modification of their virulence potential, suggesting that balance between resistance and virulence is dependent on the type of resistance mechanisms. In conclusion, this study shows that the expression of both efflux systems and porins is a key factor not only for antibiotic resistance but also virulence potential in K. pneumoniae.