EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antibacterial activities and modes of action of vancomycin and related glycopeptides.
PMID 2069366
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SBR00001
Vancomycin hydrochloride, Ready Made Solution, 100 mg/mL in DMSO, 0.2 μm filtered
C66H75Cl2N9O24 · xHCl