EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Structure and synthesis of an immunoactive lipopeptide, WS1279, of microbial origin.
PMID 2070441
Abstract
The structure of WS1279, isolated from Streptomyces sp. as an immunoactive lipopeptide, has been deduced on the basis of chemical and physical evidence as S-[2,3-bis(palmitoyloxy)propyl]-N alpha-palmitoyl-Cys-Asn-Ser-Gly-Gly-Ser- OH. This was confirmed by synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20589
O-tert-Butyl-L-serine tert-butyl ester hydrochloride, ≥98.0% (AT)
C11H23NO3 · HCl