EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Chiral mixed secondary phosphine-oxide-phosphines: high-performing and easily accessible ligands for asymmetric hydrogenation.
PMID 20715032