EMAIL THIS PAGE TO A FRIEND
Journal of cellular and molecular medicine
Role of guanylate binding protein-1 in vascular defects associated with chronic inflammatory diseases.
PMID 20716116
Abstract
Rheumatic autoimmune disorders are characterized by a sustained pro-inflammatory microenvironment associated with impaired function of endothelial progenitor cells (EPC) and concomitant vascular defects. Guanylate binding protein-1 (GBP-1) is a marker and intracellular regulator of the inhibition of proliferation, migration and invasion of endothelial cells induced by several pro-inflammatory cytokines. In addition, GBP-1 is actively secreted by endothelial cells. In this study, significantly increased levels of GBP-1 were detected in the sera of patients with chronic inflammatory disorders. Accordingly we investigated the function of GBP-1 in EPC. Interestingly, stable expression of GBP-1 in T17b EPC induced premature differentiation of these cells, as indicated by a robust up-regulation of both Flk-1 and von Willebrand factor expression. In addition, GBP-1 inhibited the proliferation and migration of EPC in vitro. We confirmed that GBP-1 inhibited vessel-directed migration of EPC at the tissue level using the rat arterio-venous loop model as a novel quantitative in vivo migration assay. Overall, our findings indicate that GBP-1 contributes to vascular dysfunction in chronic inflammatory diseases by inhibiting EPC angiogenic activity via the induction of premature EPC differentiation.