EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Nitric oxide amplifies the rat electroretinogram.
PMID 20732319
Abstract
It is well established that nitric oxide (NO) participates in retinal signal processing through stimulation ofxa0its receptor enzyme, soluble guanylyl cyclase (sGC). However, under pathological conditions such as uveoretinitis, diabetic or ischemic retinopathy, elevated NO concentrations may cause protein S-nitrosation and peroxynitrite formation in the retina, promoting cellular injury and apoptosis. Previous electroretinogram (ERG) studies demonstrated deleterious effects of NO on the retinal light response, but showed no evidence for a role in normal signal processing. To better understand the function of NO inxa0ocular physiology, we investigated the effects of exogenous NO, produced by NO donors with different release kinetics, on the flash ERG of the rat. Within a limited concentration range, NO strongly amplified ERG a- and b-waves, oscillatory potentials, and the scotopic threshold response. Amplification exceeded 100% under dark adaptation, whereas the photopic ERG and the isolated cone response were increased by less than 50%. Blocking photoreceptor-bipolar cell synapses by AP-4 demonstrated a significant increase of the isolated a-wave by NO, and modeling the ERG generator PIII supported photoreceptors as primary NO targets. The sGC inhibitors ODQ and NS2028 did not reduce NO-dependent ERG amplification, ruling outxa0an involvement of the classical NO effector cyclic GMP. Using immunohistochemistry, we show that illumination and exogenous NO altered the S-nitrosation level of the photoreceptor layer, suggesting that direct protein modifications caused by elevated levels of NO may be responsible for the observed phenomenon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3636
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
C9H5N3O2
M1555
MAHMA NONOate
C8H20N4O2